6-Chloroquinazolin-4-amine 98%

Supplier: Ambeed
Warning

A344437-100G A344437-100MG A344437-1G A344437-250MG A344437-25G A344437-5G
76773-216EA 1154.26 USD
76773-216 76773-218 76773-220 76773-222 76773-224 76773-226
6-Chloroquinazolin-4-amine 98%
6-Chloroquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₆ClN₃
MW: 179.6063 g/mol
MDL Number: MFCD02676502
CAS Number: 19808-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR