5-(3-Bromophenyl)isoxazole 97%

Supplier: Ambeed
Danger

A343572-100MG A343572-1G A343572-250MG A343572-5G
76773-194EA 29.71 USD
76773-194 76773-196 76773-198 76773-200
5-(3-Bromophenyl)isoxazole 97%
5-(3-Bromophenyl)isoxazole

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.06 g/mol
MDL Number: MFCD02183531
CAS Number: 7064-33-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR