4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine 2-hydroxysuccinate 98%

Supplier: Ambeed
Warning

Synonyms: Pizotifen malate

A343260-100MG A343260-1MG A343260-250MG
76773-188EA 9.55 USD
76773-188 76773-190 76773-192
4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine 2-hydroxysuccinate 98%
4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine 2-hydroxysuccinate

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅S
MW: 429.53 g/mol
MDL Number: MFCD01700978
CAS Number: 5189-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR