2,3,4-Trifluorobenzaldehyde 96%

Supplier: Ambeed
Danger

A342572-10G A342572-1G A342572-25G A342572-5G
76773-168EA 31.83 USD
76773-168 76773-170 76773-172 76773-174
2,3,4-Trifluorobenzaldehyde 96%
2,3,4-Trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₃O
MW: 160.1 g/mol
MDL Number: MFCD00061230
CAS Number: 161793-17-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR