2-Chloro-5-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-nitrobenzotrifluoride, 2-Chloro-α,α,α-trifluoro-5-nitrotoluene, 1-Chloro-4-nitro-2-(trifluoromethyl)benzene

A342085-100G A342085-500G A342085-25G
76773-150EA 12 USD
76773-150 76773-152 76773-154 76839-868
2-Chloro-5-nitrobenzotrifluoride 98%
2-Chloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MDL Number: MFCD00007296
CAS Number: 777-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR