5,8-Dibromoquinoline 98%

Supplier: Ambeed
Warning

A341493-1G A341493-5G
76773-142EA 11 USD
76773-142 76773-144
5,8-Dibromoquinoline 98%
5,8-Dibromoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Br₂N
MDL Number: MFCD04966994
CAS Number: 81278-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR