4-Ethoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

A341212-10G A341212-1G A341212-250MG A341212-25G A341212-5G
76773-126EA 69 USD
76773-126 76773-128 76773-130 76773-132 76773-134
4-Ethoxy-3-nitropyridine 97%
4-Ethoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD00234959
CAS Number: 1796-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR