Acrylodan 97%

Supplier: Ambeed
Warning

Synonyms: 1-(6-(Dimethylamino)naphthalen-2-yl)prop-2-en-1-one

A338441-100MG A338441-250MG A338441-1G A338441-50MG
76773-120EA 189 USD
76773-120 76773-122 77878-039 77878-040
Acrylodan 97%
Acrylodan

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO
MDL Number: MFCD00467545
CAS Number: 86636-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR