3-Fluoro-2-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A334352-1G A334352-250MG A334352-5G
76773-102EA 18 USD
76773-102 76773-104 76773-106
3-Fluoro-2-(trifluoromethyl)benzonitrile 98%
3-Fluoro-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₄N
MDL Number: MFCD01631566
CAS Number: 261951-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR