6-Chloro-N,2-dimethyl-4-pyrimidinamine 97%

Supplier: Ambeed
Warning

A333689-1G A333689-5G
76773-098EA 39.26 USD
76773-098 76773-100
6-Chloro-N,2-dimethyl-4-pyrimidinamine 97%
6-Chloro-N,2-dimethyl-4-pyrimidinamine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.6 g/mol
MDL Number: MFCD09972247
CAS Number: 5621-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR