5-Fluoro-2-methylbenzamide 97+%

Supplier: Ambeed
Warning

A332152-1G A332152-250MG A332152-5G
76773-088EA 28.99 USD
76773-088 76773-090 76773-092
5-Fluoro-2-methylbenzamide 97+%
5-Fluoro-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO
MDL Number: MFCD00205105
CAS Number: 175278-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR