2-(Chloromethyl)quinazolin-4(3H)-one 97%

Supplier: Ambeed
Danger

A331927-100MG A331927-1G A331927-250MG A331927-5G
76773-080EA 12 USD
76773-080 76773-082 76773-084 76773-086
2-(Chloromethyl)quinazolin-4(3H)-one 97%
2-(Chloromethyl)quinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MW: 194.618 g/mol
MDL Number: MFCD01927196
CAS Number: 3817-05-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR