trans-2,3-Dibromo-2-butene-1,4-diol 97%

Supplier: Ambeed
Warning

A329601-100G A329601-25G
76773-066EA 10.61 USD
76773-066 76773-068
trans-2,3-Dibromo-2-butene-1,4-diol 97%
trans-2,3-Dibromo-2-butene-1,4-diol

Precaución: for Research Use Only

Formula: C₄H₆Br₂O₂
MW: 245.9 g/mol
MDL Number: MFCD00004698
CAS Number: 21285-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR