4-Fluoro-2-methoxybenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A328033-100G
A328033-10G
A328033-1G
A328033-25G
A328033-5G
76773-052EA
424.36
USD
76773-052
76773-054
76773-056
76773-058
76773-060
4-Fluoro-2-methoxybenzonitrile 97%
4-Fluoro-2-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆FNO MW: 151.14 g/mol |
MDL Number:
MFCD04116335 CAS Number: 191014-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...