6-Fluoro-1H-benzimidazole-2-thiol 95%

Supplier: Ambeed
Warning

A327587-1G A327587-250MG A327587-25G A327587-5G
76773-036EA 19 USD
76773-036 76773-038 76773-040 76773-042
6-Fluoro-1H-benzimidazole-2-thiol 95%
6-Fluoro-1H-benzimidazole-2-thiol

Precaución: for Research Use Only

Formula: C₇H₅FN₂S
MDL Number: MFCD07432806
CAS Number: 583-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR