2-Chloro-6-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-hydroxybenzoic acid

A327508-100G A327508-10G A327508-1G A327508-250MG A327508-25G A327508-5G
76773-024EA 1078 USD
76773-024 76773-026 76773-028 76773-030 76773-032 76773-034
2-Chloro-6-hydroxybenzoic acid 98%
2-Chloro-6-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
MDL Number: MFCD10687031
CAS Number: 56961-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR