2-Fluoro-5-(trifluoromethyl)benzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-fluoro-5-(trifluoromethyl)benzene, 2-Fluoro-5-(trifluoromethyl)benzyl Bromide

A326635-10G A326635-1G A326635-25G A326635-5G
76772-994EA 63 USD
76772-994 76772-996 76772-998 76773-000
2-Fluoro-5-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-5-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrF₄
MW: 257.03
Boiling Pt: 197
Melting Pt: 22
MDL Number: MFCD00061174
CAS Number: 220239-69-0
UN: 1760
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR