5-Bromobenzoxazole-2-thiol 95%

Supplier: Ambeed
Warning

A324417-1G A324417-250MG A324417-5G
76772-970EA 27 USD
76772-970 76772-972 77626-602
5-Bromobenzoxazole-2-thiol 95%
5-Bromobenzoxazole-2-thiol

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
MDL Number: MFCD08447020
CAS Number: 439607-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR