D-ɑ-Tocopherol succinate 98%

Supplier: Ambeed
Warning

Synonyms: Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid

A324405-25G A324405-500G A324405-100G
76772-966EA 14 USD
76772-966 76772-968 77626-600
D-ɑ-Tocopherol succinate 98%
D-alpha-Tocopherol succinate

Precaución: for Research Use Only

Formula: C₃₃H₅₄O₅
MW: 530.79 g/mol
MDL Number: MFCD00072055
CAS Number: 4345-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR