5-Chloro-2-(trichloromethyl)benzimidazole 95%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-2-(trichloromethyl)-1H-benzo[d]imidazole

A323296-100MG A323296-1G A323296-250MG
76772-960EA 25.47 USD
76772-960 76772-962 76772-964
5-Chloro-2-(trichloromethyl)benzimidazole 95%
5-Chloro-2-(trichloromethyl)benzimidazole

Precaución: for Research Use Only

Formula: C₈H₄Cl₄N₂
MW: 269.934 g/mol
MDL Number: MFCD00005595
CAS Number: 3584-66-5
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR