3,5-Dichlorophenyl isocyanate 95%

Supplier: Ambeed
Danger

A322940-1G A322940-25G A322940-5G
76772-952EA 13 USD
76772-952 76772-954 77878-020
3,5-Dichlorophenyl isocyanate 95%
3,5-Dichlorophenyl isocyanate

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD00013859
CAS Number: 34893-92-0
UN: 2206
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR