Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: (2,3,4,5-Tetrafluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionate, Ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate, 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionic Acid Ethyl Ester

A322769-100G A322769-5G
76772-950EA 115.64 USD
76772-950 77571-910
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate 97%
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₈F₄O₃
MW: 264.18 g/mol
MDL Number: MFCD00523020
CAS Number: 94695-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR