5-Methylthiophene-2-carbonyl chloride 95%

Supplier: Ambeed
Danger

A322472-1G A322472-250MG
76772-940EA 131 USD
76772-940 76772-942
5-Methylthiophene-2-carbonyl chloride 95%
5-Methylthiophene-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C₆H₅ClOS
MDL Number: MFCD00130092
CAS Number: 31555-59-6
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR