4-Bromo-2-Fluorobenzylamine 98%

Supplier: Ambeed
Danger

A322237-10G A322237-5G A322237-1G
76772-936EA 65 USD
76772-936 76772-938 77878-017
4-Bromo-2-Fluorobenzylamine 98%
4-Bromo-2-Fluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD00153076
CAS Number: 112734-22-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR