4-Methylphenylboronic acid neopentyl glycol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylphenylboronic acid neopentyl ester, 5,5-Dimethyl-2-(p-tolyl)-1,3,2-dioxaborinane, 4-Methylbenzeneboronic acid neopentyl glycol ester

A321895-1G A321895-25G A321895-5G
76772-930EA 30.77 USD
76772-930 76772-932 76772-934
4-Methylphenylboronic acid neopentyl glycol ester 98%
4-Methylphenylboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₂
MW: 204.08 g/mol
MDL Number: MFCD03788728
CAS Number: 380481-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR