5,7-dihydroindolo[2,3-b]carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroindolo[2,3-b]carbazole

A321536-5G A321536-250MG A321536-25G
76772-926EA 270.53 USD
76772-926 77593-910 77626-592
5,7-dihydroindolo[2,3-b]carbazole 98%
5,7-dihydroindolo[2,3-b]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂
MW: 256.308 g/mol
MDL Number: MFCD10687177
CAS Number: 111296-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR