5-Chloro-1-methyl-1H-benzo[d][1,3]oxazine-2,4-dione 97%

Supplier: Ambeed
Warning

A321146-10G A321146-1G A321146-250MG A321146-5G
76772-918EA 301.3 USD
76772-918 76772-920 76772-922 76772-924
5-Chloro-1-methyl-1H-benzo[d][1,3]oxazine-2,4-dione 97%
5-Chloro-1-methyl-1H-benzo[d][1,3]oxazine-2,4-dione

Precaución: for Research Use Only

Formula: C₉H₆ClNO₃
MW: 211.60 g/mol
MDL Number: MFCD00047630
CAS Number: 40707-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR