6-Bromo-2-methylbenzo[d]oxazole 97%

Supplier: Ambeed
Warning

A319846-1G A319846-250MG A319846-5G
76772-910EA 125.19 USD
76772-910 76772-912 76772-914 76772-916
6-Bromo-2-methylbenzo[d]oxazole 97%
6-Bromo-2-methylbenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD06659628
CAS Number: 151230-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR