(2,4-Dichlorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A319562-100G A319562-10G A319562-25G A319562-500G
76772-898EA 12 USD
76772-898 76772-900 76772-902 76772-904
(2,4-Dichlorophenyl)acetic acid 98%
(2,4-Dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)CH₂COOH
MDL Number: MFCD00004318
CAS Number: 19719-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR