4-Chloro-2-fluoro-5-sulfamoylbenzoic acid 98%

Supplier: Ambeed
Warning

A319085-250MG A319085-1G
76772-896EA 7 USD
76772-896 77598-416
4-Chloro-2-fluoro-5-sulfamoylbenzoic acid 98%
4-Chloro-2-fluoro-5-sulfamoylbenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₄S
MDL Number: MFCD03265327
CAS Number: 4793-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR