Indolin-5-amine dihydrochloride 95%

Supplier: Ambeed
Danger

A317399-250MG A317399-100MG A317399-1G
76772-870EA 43 USD
76772-870 77878-009 77878-010
Indolin-5-amine dihydrochloride 95%
Indolin-5-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂
MW: 207.10 g/mol
MDL Number: MFCD01310783
CAS Number: 2759-14-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR