2-Chloro-5-fluorobenzonitrile 98%
Supplier: Ambeed
|
Danger
|
A317148-1G
A317148-5G
A317148-25G
76772-862EA
10
USD
76772-862
76772-864
77878-008
2-Chloro-5-fluorobenzonitrile 98%
2-Chloro-5-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃ClFN |
MDL Number:
MFCD03094168 CAS Number: 57381-56-3 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...