DDe-OH 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Hydroxyethylidene)-5,5-dimethylcyclohexane-1,3-dione

A316696-1G A316696-25G A316696-5G A316696-100G
76772-854EA 15 USD
76772-854 76772-856 76772-858 77878-006
DDe-OH 98%
DDe-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD03001669
CAS Number: 94142-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR