6,6'-Dibromo-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

Synonyms: 6,6'-Dibromo-2,2'-bipyridine

A316650-1G A316650-250MG A316650-25G A316650-5G A316650-100G A316650-100MG
76772-846EA 102.91 USD
76772-846 76772-848 76772-850 76772-852 77626-574 77626-576
6,6'-Dibromo-2,2'-bipyridyl 97%
6,6'-Dibromo-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂N₂
MW: 313.98 g/mol
MDL Number: MFCD00234587
CAS Number: 49669-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR