5-Bromo-3-nitrosalicylaldehyde 97%
Supplier: Ambeed
|
Danger
|
A314836-10G
A314836-1G
A314836-250MG
A314836-25G
A314836-5G
A314836-100G
76772-834EA
58
USD
76772-834
76772-836
76772-838
76772-840
76772-842
77626-570
5-Bromo-3-nitrosalicylaldehyde 97%
5-Bromo-3-nitrosalicylaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₄ |
MDL Number:
MFCD00130110 CAS Number: 16634-88-1 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...