5-Chloro-2-methylpyrimidine 95%

Supplier: Ambeed
Warning

A314304-100MG A314304-250MG A314304-5G A314304-1G
76772-828EA 22 USD
76772-828 76772-830 76772-832 77593-876
5-Chloro-2-methylpyrimidine 95%
5-Chloro-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C5H5ClN2
MDL Number: MFCD10574686
CAS Number: 54198-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR