5-Fluoro-2-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoro-2-methylphenyl)methanol

A313375-1G A313375-5G
76772-816EA 16 USD
76772-816 76772-818
5-Fluoro-2-methylbenzyl alcohol 97%
5-Fluoro-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16
MDL Number: MFCD01631527
CAS Number: 22062-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR