Pizotifen 98%

Supplier: Ambeed
Warning

Synonyms: 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran, 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine

A312001-250MG A312001-5G
76772-798EA 108.22 USD
76772-798 76772-800
Pizotifen 98%
Pizotifen

Precaución: for Research Use Only

Formula: C₁₉H₂₁NS
MW: 295.44 g/mol
MDL Number: MFCD00864192
CAS Number: 15574-96-6
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR