5-Chlorobenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A310983-1G A310983-250MG
76772-784EA 29.71 USD
76772-784 76772-786
5-Chlorobenzo[d]thiazole 95%
5-Chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MW: 169.63 g/mol
MDL Number: MFCD00227392
CAS Number: 2786-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR