(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-1,2,3,4-Tetrahydronaphthoic acid

A310787-10G A310787-1G A310787-250MG A310787-25G A310787-5G
76772-768EA 331.01 USD
76772-768 76772-770 76772-772 76772-774 76772-776
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD07368364
CAS Number: 23357-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR