3',4'-Dimethoxy-2,2,2-trifluoroacetophenone 95%

Supplier: Ambeed
Warning

A310116-250MG A310116-1G A310116-5G
76772-758EA 40 USD
76772-758 77626-548 77626-550
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone 95%
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MDL Number: MFCD01319991
CAS Number: 300374-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR