Tetradecyltrimethylammonium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
TTAC, Tetradonium chloride, N,N,N-Trimethyltetradecan-1-aminium chloride, Trimethyl-tetradecylammonium chloride, Myristyltrimethylammonium chloride, n-Tetradecyltrimethylammonium chloride, (1-Tetradecyl)trimethylammonium chloride
A310088-10G
A310088-1000G
A310088-100G
A310088-500G
76772-756EA
5
USD
76772-756
77598-408
77598-410
77598-412
Tetradecyltrimethylammonium chloride 97%
Tetradecyltrimethylammonium chloride
Precaución: for Research Use Only
Formula:
C₁₇H₃₈ClN MW: 291.95 g/mol |
MDL Number:
MFCD00059968 CAS Number: 4574-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...