2-(5'-Hexyl-[2,2'-bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A306880-100MG A306880-1G A306880-250MG A306880-5G
76772-728EA 33 USD
76772-728 76772-730 76772-732 77877-983
2-(5'-Hexyl-[2,2'-bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5'-Hexyl-[2,2'-bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₀H₂₉BO₂S₂
MDL Number: MFCD05664380
CAS Number: 579503-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR