5-Fluorobenzo[b]thiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A304316-1G A304316-25G A304316-5G
76772-678EA 28 USD
76772-678 76772-680 76772-682
5-Fluorobenzo[b]thiophene-2-carboxylic acid 95%
5-Fluorobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₅FO₂S
MDL Number: MFCD01927184
CAS Number: 70060-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR