5-Methylnicotinaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-3-pyridinecarboxaldehyde

A301786-1G A301786-250MG A301786-5G
76772-664EA 110.34 USD
76772-664 76772-666 76772-668
5-Methylnicotinaldehyde 96%
5-Methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C7H7NO
MW: 121.1366 g/mol
MDL Number: MFCD08272078
CAS Number: 100910-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR