Hept-6-enoic acid 98% stabilized
Supplier: Ambeed
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A301413-1G
A301413-25G
A301413-5G
76772-658EA
8
USD
76772-658
77550-826
77550-828
Hept-6-enoic acid 98% stabilized
Hept-6-enoic acid
Precaución: for Research Use Only
Formula:
C₇H₁₂O₂ |
MDL Number:
MFCD00014382 CAS Number: 1119-60-4 UN: 3265 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...