4-Fluoro-2-nitrotoluene 98%
Supplier: Ambeed
|
Warning
|
A301340-1000G
A301340-100G
A301340-10G
A301340-25G
A301340-500G
A301340-5G
76772-646EA
147
USD
76772-646
76772-648
76772-650
76772-652
76772-654
76772-656
4-Fluoro-2-nitrotoluene 98%
4-Fluoro-2-nitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₆FNO₂ |
MDL Number:
MFCD00007200 CAS Number: 446-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...