5-Chloro-2-indanone 95%

Supplier: Ambeed
Warning

A300687-1G A300687-250MG A300687-5G
76772-622EA 84 USD
76772-622 76772-624 76772-626
5-Chloro-2-indanone 95%
5-Chloro-2-indanone

Precaución: for Research Use Only

Formula: C₉H₇ClO
MDL Number: MFCD09744099
CAS Number: 74444-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR