Ethyl-5-cyanoindole-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-cyanoindole-2-carboxylate
A300143-1G
A300143-250MG
A300143-5G
A300143-10G
76772-618EA
24
USD
76772-618
76772-620
77598-404
77877-972
Ethyl-5-cyanoindole-2-carboxylate 98%
Ethyl-5-cyanoindole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀N₂O₂ |
MDL Number:
MFCD04966999 CAS Number: 105191-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...