2,3,5-Trifluorobenzaldehyde 97%

Supplier: Ambeed
Danger

A299162-10G A299162-1G A299162-25G A299162-5G
76772-604EA 141.1 USD
76772-604 76772-606 76772-608 76772-610
2,3,5-Trifluorobenzaldehyde 97%
2,3,5-Trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₃O
MW: 160.1 g/mol
MDL Number: MFCD00083538
CAS Number: 126202-23-1
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR